accessibility menu, dialog, popup

UserName

CH3H2NNH2CH3

CAS RN 119-93-7

IUPAC Name: 3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diamine
Synonyms: tolidine orthotolidine 3,3'-tolidine 2-tolidine tetrahydrofluoride diaminoditolyl bianisidine o-tolidine 2-tolidine 2-tolidina 2-tolidine dihydrofluoride
Molecular Weight (g/mol): 212.3
Molecular Formula: C14H16N2
InChi Key: NUIURNJTPRWVAP-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1N)C1=CC=C(N)C(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (4)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (4)
  • (4)
  • (5)
  • (3)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (3)
  • (4)