accessibility menu, dialog, popup

UserName

O

CAS RN 119-61-9

IUPAC Name: diphenylmethanone
Synonyms: benzophenone benzophenone (substance) α-oxoditane epa pesticide chemical code 000315 diphenyl methanone caswell no. 081G ccris 629 Ph2CO α-oxodiphenylmethane
Molecular Weight (g/mol): 182.22
Molecular Formula: C13H10O
InChi Key: RWCCWEUUXYIKHB-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (5)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Grade 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)