accessibility menu, dialog, popup

UserName

CAS RN 119-60-8

O

CAS RN 119-60-8

IUPAC Name: dicyclohexylmethanone
Synonyms: methanone, dicyclohexyl-
Molecular Weight (g/mol): 194.32
Molecular Formula: C13H22O
InChi Key: TWXWPPKDQOWNSX-UHFFFAOYSA-N
SMILES: O=C(C1CCCCC1)C1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results