accessibility menu, dialog, popup

UserName

CAS RN 119-42-6

IUPAC Name: 2-cyclohexylphenol
Synonyms: 2-cyclohexylphenol
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: MVRPPTGLVPEMPI-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results