accessibility menu, dialog, popup

UserName

OHOO

CAS RN 118-55-8

IUPAC Name: phenyl 2-hydroxybenzoate
Synonyms: salol phenyl salicylate (product) phenyl salicylate (substance) salphenyl fenylester kyseliny salicylove salol preparation musol phenyl salicylate preparation
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: ZQBAKBUEJOMQEX-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)OC1=CC=CC=C1
Boiling Point 
  • (6)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (6)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (6)
  • (1)
  • (6)
  • (2)
  • (4)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (6)
  • (2)
  • (1)
  • (6)