accessibility menu, dialog, popup

UserName

CAS RN 117976-90-6

IUPAC Name: sodium 2-{[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methanesulfinyl}-1H-1,3-benzodiazol-1-ide
Synonyms: AcipHex aciphex sodium rabeprazole pariet
Molecular Weight (g/mol): 381.43
Molecular Formula: C18H20N3NaO3S
InChi Key: KRCQSTCYZUOBHN-UHFFFAOYNA-N
SMILES: [Na+].COCCCOC1=CC=NC(CS(=O)C2=NC3=CC=CC=C3[N-]2)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results