accessibility menu, dialog, popup

UserName

CAS RN 1176202-63-3

IUPAC Name: 1,3-bis(adamantan-1-yl)imidazolidin-1-ium; tetrafluoroboranuide
Synonyms: 1,3-bis(adamantan-1-yl)imidazolidin-1-ium tetrafluoroborate
Molecular Weight (g/mol): 428.37
Molecular Formula: C23H37BF4N2
InChi Key: AFLPPSBVXPAHAH-UHFFFAOYNA-O
SMILES: F[B-](F)(F)F.C1C[NH+](CN1C12CC3CC(CC(C3)C1)C2)C12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results