accessibility menu, dialog, popup

UserName

CAS RN 117483-89-3

IUPAC Name: 1-[(benzyloxy)carbonyl]-2,3-dihydro-1H-indole-2-carboxylic acid
Synonyms: 1-[(benzyloxy)carbonyl]-2,3-dihydroindole-2-carboxylic acid
Molecular Weight (g/mol): 297.31
Molecular Formula: C17H15NO4
InChi Key: BSOYWTITVKXHLM-UHFFFAOYNA-N
SMILES: OC(=O)C1CC2=CC=CC=C2N1C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)