accessibility menu, dialog, popup

UserName

NHNOOOHOHO(E/Z)

CAS RN 1160927-48-9

IUPAC Name: 2-{[(6-carboxyhexanamido)imino]methyl}benzoic acid
Synonyms: 2-{[(6-carboxyhexanamido)imino]methyl}benzoic acid
Molecular Weight (g/mol): 306.32
Molecular Formula: C15H18N2O5
InChi Key: ABKJCDILEUEJSH-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCC(=O)NN=CC1=CC=CC=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results