CAS RN 1156-51-0
IUPAC Name:
4-{2-[4-(cyanooxy)phenyl]propan-2-yl}phenyl cyanate
Synonyms:
4-{2-[4-(cyanooxy)phenyl]propan-2-yl}phenyl cyanate
Molecular Weight (g/mol):
278.31
Molecular Formula:
C17H14N2O2
InChi Key:
AHZMUXQJTGRNHT-UHFFFAOYSA-N
SMILES:
CC(C)(C1=CC=C(OC#N)C=C1)C1=CC=C(OC#N)C=C1
1
–
1
of
1
results
1
2,2-Bis(4-cyanatophenyl)propane 98.0+%, TCI America™
CAS: 1156-51-0 Molecular Formula: C17H14N2O2 Molecular Weight (g/mol): 278.311 MDL Number: MFCD00037423 InChI Key: AHZMUXQJTGRNHT-UHFFFAOYSA-N Synonym: Bisphenol A Dicyanate, 4,4′C-Isopropylidenebis(cyanatobenzene) PubChem CID: 70872 IUPAC Name: [4-[2-(4-cyanatophenyl)propan-2-yl]phenyl] cyanate SMILES: CC(C)(C1=CC=C(C=C1)OC#N)C2=CC=C(C=C2)OC#N