missing translation for 'onlineSavingsMsg'
Learn More

2,2-Bis(4-cyanatophenyl)propane 98.0+%, TCI America™

Catalog No. B125110G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B125110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B125110G Supplier TCI America Supplier No. B125110G

Chemical Identifiers

CAS 1156-51-0
Molecular Formula C17H14N2O2
Molecular Weight (g/mol) 278.311
MDL Number MFCD00037423
InChI Key AHZMUXQJTGRNHT-UHFFFAOYSA-N
Synonym Bisphenol A Dicyanate, 4,4′C-Isopropylidenebis(cyanatobenzene)
PubChem CID 70872
IUPAC Name [4-[2-(4-cyanatophenyl)propan-2-yl]phenyl] cyanate
SMILES CC(C)(C1=CC=C(C=C1)OC#N)C2=CC=C(C=C2)OC#N

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 10 g
Formula Weight 278.31
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Bis(4-cyanatophenyl)propane
EINECSNumber (4)-0919
TSCA No