accessibility menu, dialog, popup

UserName

CAS RN 115256-11-6

IUPAC Name: N-[4-(2-{[2-(4-methanesulfonamidophenyl)ethyl](methyl)amino}ethoxy)phenyl]methanesulfonamide
Synonyms: dofetilide dofetilide (product) dofetilida [inn-spanish] dofetilida tikosyn dofetilide (substance) 多非利特 dofetilidum 1-mspmpe dofetilide preparation
Molecular Weight (g/mol): 441.56
Molecular Formula: C19H27N3O5S2
InChi Key: IXTMWRCNAAVVAI-UHFFFAOYSA-N
SMILES: CN(CCOC1=CC=C(NS(C)(=O)=O)C=C1)CCC1=CC=C(NS(C)(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)