accessibility menu, dialog, popup

UserName

ONONNOO

CAS RN 1151-97-9

IUPAC Name: 2-[(2,4-dinitrophenyl)methyl]pyridine
Synonyms: 2-[(2,4-dinitrophenyl)methyl]pyridine
Molecular Weight (g/mol): 259.22
Molecular Formula: C12H9N3O4
InChi Key: KKFNJVINGIUTIH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CC2=CC=CC=N2)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)