accessibility menu, dialog, popup

UserName

OHOHOSOOHOHO

CAS RN 115-41-3

IUPAC Name: 3,3-bis(3,4-dihydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: 3,3-bis(3,4-dihydroxyphenyl)-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Weight (g/mol): 386.37
Molecular Formula: C19H14O7S
InChi Key: RRRCKIRSVQAAAS-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1
Quantity 
  • (2)
  • (2)
  • (3)
Grade 
  • (2)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)