missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrocatechol Violet, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 386.37 |
| Chemical Name or Material | Pyrocatechol Violet |
Safety and Handling
| TSCA | Yes |