missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Pyrocatechol Violet, pure, indicator grade
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Lambda Max | 441 to 449nm (in methanol) |
| Packaging | Glass bottle |
| Specific Extinction | 375 (at lambda max, methanol) min. |
| Suitability | for complexometric det. of metal oxides: passes |
| Color | Brown or Green |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol, 2-methoxyethanol |
| Quantity | 25 g |
| Physical Form | Metallic or Crystalline Powder |
| Formula Weight | 386.36 |
| Show More |
RUO – Research Use Only