accessibility menu, dialog, popup

UserName

BOOCH3H3CCH3CH3(?)

CAS RN 1149804-35-2

IUPAC Name: 4,4,5,5-tetramethyl-2-[10-(naphthalen-1-yl)anthracen-9-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[10-(naphthalen-1-yl)anthracen-9-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 430.35
Molecular Formula: C30H27BO2
InChi Key: HZMCBFYBIMINFR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results