accessibility menu, dialog, popup

UserName

CAS RN 1141-38-4

IUPAC Name: naphthalene-2,6-dicarboxylic acid
Synonyms: 2,6-dicarboxynaphthalene 2,6-naphthalic acid
Molecular Weight (g/mol): 216.19
Molecular Formula: C12H8O4
InChi Key: RXOHFPCZGPKIRD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC=C(C=C2C=C1)C(O)=O
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)