missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2,6-dicarboxylic acid, 98+%

Catalog No. AAA1686822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1686822 100 g
AAA1686806 5 g
AAA1686814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1686822 Supplier Thermo Scientific Chemicals Supplier No. A1686822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1141-38-4 | C12H8O4 | 216.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1141-38-4
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.192
MDL Number MFCD00004105
InChI Key RXOHFPCZGPKIRD-UHFFFAOYSA-N
Synonym 2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711
PubChem CID 14357
ChEBI CHEBI:44460
IUPAC Name naphthalene-2,6-dicarboxylic acid
SMILES C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 100 g
Beilstein 2051257
Formula Weight 216.19
Percent Purity ≥98%
Chemical Name or Material Naphthalene-2,6-dicarboxylic acid
EINECSNumber 214-527-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only