missing translation for 'onlineSavingsMsg'
Learn More

2,6-Naphthalenedicarboxylic Acid 98.0+%, TCI America™

Catalog No. N03775G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03775G 5 g
N037725G 25 g
N0377100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N03775G Supplier TCI America Supplier No. N03775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1141-38-4
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.192
MDL Number MFCD00004105
InChI Key RXOHFPCZGPKIRD-UHFFFAOYSA-N
Synonym 2,6-naphthalenedicarboxylic acid, 2,6-naphthalic acid, 2,6-dicarboxynaphthalene, unii-k3c4dyz29o, 2,6-naphthalene dicarboxylic acid, k3c4dyz29o, dsstox_cid_9211, acmc-2099kj, d06kmp, dsstox_rid_78711
PubChem CID 14357
ChEBI CHEBI:44460
IUPAC Name naphthalene-2,6-dicarboxylic acid
SMILES C1=CC2=C(C=CC(=C2)C(=O)O)C=C1C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 216.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Naphthalenedicarboxylic Acid
EINECSNumber (4)-0402
TSCA Yes