accessibility menu, dialog, popup

UserName

H3COHNNH

CAS RN 114-83-0

IUPAC Name: N'-phenylacetohydrazide
Synonyms: N(1)-acetylphenylhydrazine
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: UICBCXONCUFSOI-UHFFFAOYSA-N
SMILES: CC(=O)NNC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)