accessibility menu, dialog, popup

UserName

CAS RN 113841-59-1

IUPAC Name: 4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
Synonyms: 4-(bromomethyl)-3-methyl-5-phenyl-1,2-oxazole
Molecular Weight (g/mol): 252.11
Molecular Formula: C11H10BrNO
InChi Key: KCGVZUOBUPWFJC-UHFFFAOYSA-N
SMILES: CC1=NOC(=C1CBr)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results