CAS RN 1134-76-5
IUPAC Name:
2-[3-(dimethylamino)propoxy]aniline
Synonyms:
2-[3-(dimethylamino)propoxy]aniline
Molecular Weight (g/mol):
194.28
Molecular Formula:
C11H18N2O
InChi Key:
XGTIDBFMHOJXSS-UHFFFAOYSA-N
SMILES:
CN(C)CCCOC1=CC=CC=C1N