accessibility menu, dialog, popup

UserName

CAS RN 113020-21-6

IUPAC Name: 1-(methoxycarbonyl)cyclopropane-1-carboxylic acid
Synonyms: 1-(methoxycarbonyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H8O4
InChi Key: IZYOHLOUZVEIOS-UHFFFAOYSA-N
SMILES: COC(=O)C1(CC1)C(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results