accessibility menu, dialog, popup

UserName

CAS RN 1129-26-6

IUPAC Name: 4-methoxybenzene-1-sulfonamide
Synonyms: P-methoxybenzenesulfonamide
Molecular Weight (g/mol): 187.21
Molecular Formula: C7H9NO3S
InChi Key: MSFQEZBRFPAFEX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)S(N)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results