accessibility menu, dialog, popup

UserName

H3CONHOHONHOO

CAS RN 1128181-23-6

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-tetradecanamidohexanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-tetradecanamidohexanoic acid
Molecular Weight (g/mol): 578.79
Molecular Formula: C35H50N2O5
InChi Key: PNXSYZSEFDFTDZ-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCC(=O)NCCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results