accessibility menu, dialog, popup

UserName

CH3ClCH3Cl

CAS RN 1124-05-6

IUPAC Name: 1,4-dichloro-2,5-dimethylbenzene
Synonyms: 1,4-dichloro-2,5-dimethylbenzene
Molecular Weight (g/mol): 175.05
Molecular Formula: C8H8Cl2
InChi Key: UTGSRNVBAFCOEU-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C(C)C=C1Cl
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)