missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-p-xylene, 98%

Catalog No. AAB2134422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2134422 100 g
AAB2134414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2134422 Supplier Thermo Scientific Chemicals Supplier No. B2134422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1124-05-6 | C8H8Cl2 | 175.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1124-05-6
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000610
InChI Key UTGSRNVBAFCOEU-UHFFFAOYSA-N
Synonym 2,5-dichloro-p-xylene, 2,5-dichloro-1,4-dimethylbenzene, benzene, 1,4-dichloro-2,5-dimethyl, p-xylene, 2,5-dichloro, unii-7p8m86zob2, 2,5-dichloro-para-xylene, p-xylene,5-dichloro, p-xylene,2,5-dichloro, acmc-2099eq, dsstox_cid_24780
PubChem CID 70755
IUPAC Name 1,4-dichloro-2,5-dimethylbenzene
SMILES CC1=CC(=C(C=C1Cl)C)Cl

Specifications

Melting Point 68°C to 70°C
Quantity 100 g
Boiling Point 221°C to 223°C
Formula Weight 175.06
Percent Purity 98%
Chemical Name or Material 2,5-Dichloro-p-xylene
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-387-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only