missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-p-xylene 97.0+%, TCI America™

Catalog No. D043125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D043125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D043125G Supplier TCI America Supplier No. D043125G

Chemical Identifiers

CAS 1124-05-6
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000610
InChI Key UTGSRNVBAFCOEU-UHFFFAOYSA-N
Synonym 2,5-dichloro-p-xylene, 2,5-dichloro-1,4-dimethylbenzene, benzene, 1,4-dichloro-2,5-dimethyl, p-xylene, 2,5-dichloro, unii-7p8m86zob2, 2,5-dichloro-para-xylene, p-xylene,5-dichloro, p-xylene,2,5-dichloro, acmc-2099eq, dsstox_cid_24780
PubChem CID 70755
IUPAC Name 1,4-dichloro-2,5-dimethylbenzene
SMILES CC1=CC(=C(C=C1Cl)C)Cl

Specifications

Melting Point 70°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 108°C
Formula Weight 175.05
Percent Purity ≥97.0% (GC)
Chemical Name or Material 2,5-Dichloro-p-xylene