accessibility menu, dialog, popup

UserName

NH2FFN

CAS RN 112279-61-5

IUPAC Name: 4-amino-2,5-difluorobenzonitrile
Synonyms: 4-amino-2,5-difluorobenzonitrile
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H4F2N2
InChi Key: LAPGMTOHOQPDGI-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(C=C1F)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)