accessibility menu, dialog, popup

UserName

IN

CAS RN 1120-90-7

IUPAC Name: 3-iodopyridine
Synonyms: 3-iodopyridine
Molecular Weight (g/mol): 205
Molecular Formula: C5H4IN
InChi Key: XDELKSRGBLWMBA-UHFFFAOYSA-N
SMILES: IC1=CC=CN=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)