accessibility menu, dialog, popup

UserName

CH3OOCH3(Z)

CAS RN 1120-34-9

IUPAC Name: methyl (13Z)-docos-13-enoate
Synonyms: methyl (13Z)-docos-13-enoate
Molecular Weight (g/mol): 352.6
Molecular Formula: C23H44O2
InChi Key: ZYNDJIBBPLNPOW-KHPPLWFESA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results