missing translation for 'onlineSavingsMsg'
Learn More

Methyl cis-13-Docosenoate 90.0+%, TCI America™

Catalog No. D1017500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1017500ML 500 mL
D101725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1017500ML Supplier TCI America Supplier No. D1017500ML

Chemical Identifiers

CAS 1120-34-9
Molecular Formula C23H44O2
Molecular Weight (g/mol) 352.603
MDL Number MFCD00027343
InChI Key ZYNDJIBBPLNPOW-KHPPLWFESA-N
Synonym methyl erucate, erucic acid methyl ester, methyl cis-13-docosenoate, z-methyl docos-13-enoate, methyl z-docos-13-enoate, methyl z-13 docosenoate, unii-dt8r52277s, 13-docosenoic acid, methyl ester, z, methyl 13z-docos-13-enoate, 13-docosenoic acid, methyl ester, 13z
PubChem CID 5364423
IUPAC Name methyl (Z)-docos-13-enoate
SMILES CCCCCCCCC=CCCCCCCCCCCCC(=O)OC

Specifications

Color Yellow
Boiling Point 222°C
Quantity 500 mL
UN Number 3272
Formula Weight 352.60
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl cis-13-Docosenoate
EINECSNumber (2)-0798
TSCA Yes