accessibility menu, dialog, popup

UserName

H3CO

CAS RN 112-31-2

IUPAC Name: decanal
Synonyms: decanal decanal (natural) N-decanal caprinic aldehyde capraldehyde 1-decanal(mixed isomers) caprinaldehyde aldehyde C10 C-10 aldehyde capric aldehyde
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: KSMVZQYAVGTKIV-UHFFFAOYSA-N
SMILES: CCCCCCCCCC=O
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (1)