accessibility menu, dialog, popup

UserName

ClH3CNCH3CH3H3C

CAS RN 112-03-8

IUPAC Name: trimethyl(octadecyl)azanium chloride
Synonyms: steartrimonium chloride
Molecular Weight (g/mol): 348.06
Molecular Formula: C21H46ClN
InChi Key: VBIIFPGSPJYLRR-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)C
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)