accessibility menu, dialog, popup

UserName

BrCH2

CAS RN 1119-51-3

IUPAC Name: 5-bromopent-1-ene
Synonyms: 1-pentene, 5-bromo-
Molecular Weight (g/mol): 149.03
Molecular Formula: C5H9Br
InChi Key: LPNANKDXVBMDKE-UHFFFAOYSA-N
SMILES: BrCCCC=C
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)