missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-pentene, 95%

Catalog No. AC295170500
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295170500 50 g Glass bottle
AC295170025 2.5 g Glass bottle
AC295170100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC295170500 Supplier Thermo Scientific Chemicals Supplier No. 295170500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1119-51-3 | C5H9Br | 149.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1119-51-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.03
MDL Number MFCD00000264
InChI Key LPNANKDXVBMDKE-UHFFFAOYSA-N
Synonym 5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide
PubChem CID 70704
IUPAC Name 5-bromopent-1-ene
SMILES C=CCCCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 1.2600g/mL
Boiling Point 126.0°C to 127.0°C (765.0 mmHg)
Flash Point 30°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.4650
Linear Formula Br(CH2)3CHCH2
Quantity 50 g
UN Number 1993
Specific Gravity 1.26
Formula Weight 149.03
Percent Purity 95%
Chemical Name or Material 5-Bromo-1-pentene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-281-4

RUO – Research Use Only