accessibility menu, dialog, popup

UserName

H3CCH3H3COOCH3CH3H3C

CAS RN 1118-71-4

IUPAC Name: 2,2,6,6-tetramethylheptane-3,5-dione
Synonyms: 2,2,6,6-tetramethylheptane-3,5-dione
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: YRAJNWYBUCUFBD-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)CC(=O)C(C)(C)C
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)