accessibility menu, dialog, popup

UserName

HOOHOHOHO

CAS RN 1114-34-7

IUPAC Name: 2,3,4,5-tetrahydroxypentanal
Synonyms: (+)-xylose L-(+)-arabinose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: PYMYPHUHKUWMLA-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(O)C=O
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (5)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)