missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Lyxose, 99+%, mixture of anomers

Catalog No. AC205230250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205230250 25 g Glass bottle
AC205230050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC205230250 Supplier Thermo Scientific Chemicals Supplier No. 205230250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1114-34-7 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1114-34-7
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064362
InChI Key PYMYPHUHKUWMLA-UOWFLXDJSA-N
Synonym 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
PubChem CID 65550
ChEBI CHEBI:16789
IUPAC Name (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 108.0°C to 112.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 31, 56
Merck Index 15, 5702
Specific Rotation Condition − 14.00 (20.00°C c=4,H2O,NH3)
Specific Rotation − 14.00
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in ethanol
Formula Weight 150.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Lyxose, Mixture of anomers
Show More Show Less
EINECSNumber 214-212-8

RUO – Research Use Only