accessibility menu, dialog, popup

UserName

CAS RN 111-88-6

IUPAC Name: octane-1-thiol
Synonyms: 1-octanethiol
Molecular Weight (g/mol): 146.29
Molecular Formula: C8H18S
InChi Key: KZCOBXFFBQJQHH-UHFFFAOYSA-N
SMILES: CCCCCCCCS
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)