accessibility menu, dialog, popup

UserName

CAS RN 111-31-9

IUPAC Name: hexane-1-thiol
Synonyms: 1-hexanethiol
Molecular Weight (g/mol): 118.24
Molecular Formula: C6H14S
InChi Key: PMBXCGGQNSVESQ-UHFFFAOYSA-N
SMILES: CCCCCCS
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)