accessibility menu, dialog, popup

UserName

CAS RN 11006-34-1

IUPAC Name: chlorophyllin
Synonyms: chlorophyllin
Molecular Weight (g/mol): 724.16
Molecular Formula: C34H31CuN4Na3O6
InChi Key: HWDGVJUIHRPKFR-UHFFFAOYNA-I
SMILES: [Na+].[Na+].[Na+].[Cu++].CCC1=C(C)C2=N/C/1=C\C1=C(C)C(=C([O-])[O-])C(=N1)\C(CC([O-])=O)=C1/N=C(\C=C3/[N-]/C(=C\2)C(C=C)=C3C)C(C)C1CCC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)