accessibility menu, dialog, popup

UserName

H3COHNHCH3OH

CAS RN 110-97-4

IUPAC Name: 1-[(2-hydroxypropyl)amino]propan-2-ol
Synonyms: diisopropanolamine
Molecular Weight (g/mol): 133.19
Molecular Formula: C6H15NO2
InChi Key: LVTYICIALWPMFW-UHFFFAOYNA-N
SMILES: CC(O)CNCC(C)O
Boiling Point 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)