missing translation for 'onlineSavingsMsg'
Learn More

Diisopropanolamine (DL- and meso- mixture) 90.0+%, TCI America™

Catalog No. D0924500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0924500G 500 g
D092425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0924500G Supplier TCI America Supplier No. D0924500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-97-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00004531
InChI Key LVTYICIALWPMFW-UHFFFAOYNA-N
Synonym diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine
PubChem CID 8086
IUPAC Name 1-[(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(O)CNCC(C)O

Specifications

Color White-Yellow
Boiling Point 249°C
Quantity 500 g
Formula Weight 133.19
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Diisopropanolamine (DL- and meso- mixture)
EINECSNumber (2)-0309
RTECSNumber UB6600000
TSCA Yes