missing translation for 'onlineSavingsMsg'
Learn More

Diisopropanolamine, 99%

Catalog No. AC407805000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC407805000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC407805000 Supplier Thermo Scientific Chemicals Supplier No. 407805000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 110-97-4 | C6H15NO2 | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-97-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00004531
InChI Key LVTYICIALWPMFW-UHFFFAOYNA-N
Synonym diisopropanolamine, 1,1'-iminodipropan-2-ol, bis 2-hydroxypropyl amine, 2-propanol, 1,1'-iminobis, bis 2-propanol amine, 1,1'-iminodi-2-propanol, dipa alcohol, di-2-propanolamine, 1,1'-azanediylbis propan-2-ol, dipropyl-2,2'-dihydroxy-amine
PubChem CID 8086
IUPAC Name 1-[(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(O)CNCC(C)O

Specifications

Melting Point 33°C
Color Colorless to Yellow
Density 1.0038g/mL
Boiling Point 249°C to 250°C
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 98.5 % min. (Total base)
Packaging Glass bottle
Refractive Index 1.4615 to 1.4635
Linear Formula (CH3CHOHCH2)2NH
Quantity 500 g
Specific Gravity 1.0038
Merck Index 15, 3214
Solubility Information Solubility in water: 870g/L (20°C)
Viscosity 84.19 mPa.s (60°C)
Formula Weight 133.19
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material Diisopropanolamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 203-820-9
RTECSNumber UB6600000
TSCA TSCA

RUO – Research Use Only