missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropanolamine, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 249°C / 480.2°F |
| Packaging | Poly Pail |
| Quantity | 500 g |
| Molecular Weight (g/mol) | 133.19 |
| Grade | Reagent |
| Physical Form | Crystals or Flakes |
| Chemical Name or Material | Diisopropanolamine |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.