accessibility menu, dialog, popup

UserName

CAS RN 110-80-5

IUPAC Name: 2-ethoxyethan-1-ol
Synonyms: 2-ethoxyethanol 2-ethoxyethanol (substance) solvulose EGEE plastiazan 60 solvid 2EE jeffersol EE bikanol E 1 ethyl cellosolve
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: ZNQVEEAIQZEUHB-UHFFFAOYSA-N
SMILES: CCOCCO
Quantity 
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (4)
  • (5)
  • (4)
  • (2)
Grade 
  • (2)
  • (3)
  • (9)
Physical Form 
  • (12)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (7)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (5)
  • (4)
  • (2)
  • (2)
  • (3)
  • (9)
  • (12)
  • (2)
  • (2)
  • (2)
  • (5)