missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monoethyl Ether, Reagent Grade, LabChem™

Catalog No. LC141604
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC141604 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC141604 Supplier LabChem Supplier No. LC141604

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
IUPAC Name 2-ethoxyethan-1-ol
SMILES CCOCCO

Specifications

Melting Point -100°C
Color Colorless
CAS Max % 100
Decomposition Information Carbon monoxide; Carbon dioxide; Peroxides
Quantity 4 L
Flash Point 44°C
Identification Passes Test
Packaging Amber Glass
Physical Form Liquid
Linear Formula C4H10O2
Viscosity 2.08mPa.s
UN Number UN1171
Boiling Point 135°C
Solubility Information Soluble in water
Formula Weight 90.12
Grade Reagent
Chemical Name or Material Ethylene Glycol Monoethyl Ether
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Toxic if inhaled.
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Avoid breathing vapors.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Contaminated work clothing must not be allowed out of the workplace.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 203-804-1
Recommended Storage Room Temperature