missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid 99.0+%, TCI America™

Catalog No. F0067500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0067500G 500 g
F006725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0067500G Supplier TCI America Supplier No. F0067500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-17-8
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00002700
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
PubChem CID 444972
ChEBI CHEBI:18012
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

Melting Point 287°C
Color White
pH 2.2 to 2.6 (1% H2O suspention)
Quantity 500 g
Formula Weight 116.07
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fumaric Acid
EINECSNumber (2)-1091
RTECSNumber LS9625000
TSCA Yes